Learn More
Cayman Chemical Morus rot Kuwnon C 10mg
SDP
Supplier:  Cayman Chemical 3342110 MG
Encompass_Preferred
A prenylated flavone; scavenges DPPH radicals (EC50 = 72.99 UG/ml); active against E. coli, S. typhimurium, S. epidermis, and S. aureus (MICs = 10, 25, 6.25, and 6.25 UG/ml, respectively); inhibits the growth of P388 mouse lymphoma cancer cells (IC50 = 14 UG/ml); improves motor coordination in a rotarod test and decreases neuronal loss in the substantia nigra pars compacta in a mouse model of MPTP-induced Parkinson's disease at 30 mg/kg
Catalog No. 50-294-6383