Learn More
Medchemexpress LLC Mosapride citrate dihydrate | 636582-62-2 | 99.6% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0189B5MG
Encompass_Preferred
Mosapride citrate dihydrate is a gastroprokinetic agent with 5-hydroxytryptamine4 receptor agonist activity, used in research for various gastrointestinal disorders. It potently inhibits Kv4.3 in a concentration-dependent manner and selectively stimulates upper GI motility in vivo.
  • Gastroprokinetic agent
  • 5-hydroxytryptamine4 receptor agonist activity
  • Used in research for gastrointestinal disorders
  • Potently inhibits Kv4.3 with an IC50 of 15.2 μM
  • Selectively stimulates upper GI motility in vivo
Catalog No. 50-004-06172