Learn More
Medchemexpress LLC HY-N2035 1mg Medchemexpress, Moslosooflavone CAS:3570-62-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN20351MG
Encompass_Preferred
Medchemexpress, HY-N2035 1mg Moslosooflavone CAS:3570-62-5 Moslosooflavone is a flavonoid isolated from Saussurea involucrata. Moslosooflavone has an anti-hypoxia and anti-inflammatory activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2859