Learn More
Selleck Chemical LLC Motesanib Diphosphate (AMG-706) 10mg 857876-30-3
SDP
Supplier:  Selleck Chemical LLC S103210MG
Encompass_Preferred
Motesanib Diphosphate (AMG-706) is a potent ATP-competitive inhibitor of VEGFR1/2/3 with IC50 of 2 nM/3 nM/6 nM, respectively; similar activity against Kit (c-Kit), ~10-fold more selective for VEGFR than PDGFR and Ret. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-620-6