Learn More
Microscope World MOTIC MICROSCOPE POLARIZING KI
SDP
Supplier:  Microscope World 1101.0006.00651
Encompass
Polarizing Kit for the National Optical 420 stereo microscope and the Motic SMZ-140 microscopes. Includes polarizer and analyzer. Intended for use with stand that has a light built into the base. Fits into 80mm stage plate well.
Catalog No. NC1809162