Learn More
Medchemexpress LLC HY-P1037 1mg Medchemexpress, Motilin (26-47), human, porcine CAS:52906-92-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP10371MG
Encompass_Preferred
Medchemexpress, HY-P1037 1mg Motilin (26-47), human, porcine CAS:52906-92-0 Motilin (26-47), human, porcine is an endogenous motilin receptor ligand with K i and EC 50 of 2.3 nM and 0.3 nM in a Chinese hamster ovary cell line. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4146