Learn More
Grainger Motor 1 1/2 1/5 HP 3 450/1 725 rpm 115V
Supplier:  Grainger 16U466
Encompass_Preferred
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B374, B43, B48L, B62, B73, 2HP and 3HP pumps. This motor face measures 7-1/2 inches by 5-1/8 inches and has a 1/2 inch diameter shaft. It features a 56J frame, 1.10 service factor, 3450 RPM and includes capacitor mounting holes. The Century motor face is designed for inground and aboveground pools and spas.
Catalog No. 50-276-8309