Learn More
Grainger Motor 2 HP 3 450 rpm 56Y 115/230V
Supplier:  Grainger 16U452
Encompass_Preferred
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, N324, and more. This motor face measures 10-1/2 inches by 10-1/2 inches and has a 1/2 inch shaft diameter. It can handle up to 3/4 HP and 3450 RPM. The motor face attaches to the pump housing using 4 bolts and comes with a new impeller and gasket. This is an OEM replacement part made to match the original pump motor.
Catalog No. 50-276-7368