Learn More
Medchemexpress LLC HY-P2048A 10mg Medchemexpress, MOTS-c(human) (acetate) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP2048A10MG
Encompass_Preferred
Medchemexpress, HY-P2048A 10mg MOTS-c(human) (acetate) CAS:0 MOTS-c(human) acetate is a mitochondrial-derived peptide. MOTS-c(human) acetate induces the accumulation of AMP analog AICAR, increases activation of AMPK and expression of its downstream GLUT4. MOTS-c(human) acetate induces glucose uptake and improves insulin sensitivity. MOTS-c(human) acetate has implications in the regulation of obesity, diabetes, exercise, and longevity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1704