Learn More
Dino Lite RK-10A Stable Tabletop Stand with Fine-focus Adjustment and Quick Release
Supplier:  Dino Lite RK10A
Encompass_Preferred
The Dino-Lite RK-10A universal stand is a stable tabletop platform that features precise fine-focus adjustment and is compatible with the complete line of Dino-Lite handheld microscope cameras. The RK-10A quick release button is helpful for fast and convenient vertical movement and fine-focus adjustment knob is especially useful at high magnification. Its adjustable horizontal arm has a 7-inch range with 360-degree rotation and the included optional arm attachment increases its effective range of motion. These features combine to provide complete command and control of microscope orientation and positioning for maximum productivity and ease of use. The RK-10A is composed of resilient stainless steel and lightweight aluminum. Optional accessories are available and sold separately.
Catalog No. NC1212582