Learn More
Electron Microscopy Sciences EUKITT Classic Mounting Medium 100 ML
SDP
Supplier:  Electron Microscopy Sciences 1532010
Encompass_Preferred
EUKITT classic was formulated for use in Automated Coverslipping machines and is considered the gold standard. Slide preparations with EUKITT Classic exhibit minimal discoloration and perfect drying without any bubbles and streaks. With an ideal viscosity (500-750 mPa's) it is a perfect formulation for all automated machines.
Supplied in a wide neck, unbreakable aluminum bottle. It is fast drying for rapid results, neutral and colorless. Features a well proven formulation and long term stability.
Catalog No. 50-192-8500