Learn More
American Research Products Inc Mouse A Disintegrin And Metall
Supplier:  American Research Products Inc CSBE08817M
Encompass
Mouse A Disintegrin And Metalloprotease 8 ADAM8 ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29226