Learn More
American Research Products Inc Mouse A disintegrin and metall
Supplier:  American Research Products Inc CSBEL001311MO
Encompass
Mouse A disintegrin and metalloproteinase with thrombospondin motifs 4 (ADAMTS4) ELISA kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/ mL Sensitivity 0 78 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28375