Learn More
American Research Products Inc Mouse adrenocorticotropic horm
Supplier:  American Research Products Inc CSBE06874M
Encompass
Mouse adrenocorticotropic hormone (ACTH) ELISA kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 68 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-28964