Learn More
American Research Products Inc Mouse adVanced glycation end p
Supplier:  American Research Products Inc CSBE09414M
Encompass
Mouse adVanced glycation end products AGEs ELISA Kit from CUSABIO Detection Range 0 156 g/mL-10 g/mL Sensitivity 0 039 g/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28699