Learn More
American Research Products Inc Mouse alpha 1 microglobulin bi
Supplier:  American Research Products Inc CSBE09776M
Encompass
Mouse alpha-1-microglobulin/bikunin precursor (AMBP) ELISA kit from CUSABIO Detection Range 1 563 ng/mL-100 ng/mL Sensitivity 0 649 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29341