Learn More
American Research Products Inc Mouse Alpha 2 macroglobulin A
Supplier:  American Research Products Inc CSBEL001004MO
Encompass
Mouse Alpha-2-macroglobulin (A2M) ELISA kit from CUSABIO Detection Range 0 156 g/mL-10 g/mL Sensitivity 0 039 g/mL Sample serum plasma Method Competitive
Catalog No. 50-003-29385