Learn More
American Research Products Inc Mouse Angiopoietin related pro
Supplier:  American Research Products Inc CSBEL001712MO
Encompass
Mouse Angiopoietin-related protein 4 (ANGPTL4) ELISA kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-27593