Learn More
Cayman Chemical Mouse Anti-6HIS IgG DyLight
SDP
Supplier:  Cayman Chemical 11117100 UG
Encompass_Preferred
Mouse anti-6His (6x Histidine) IgG (Clone AD1.1.10) conjugated to DyLight 650 Antibody: Mouse monoclonal Anti-6xHistidine IgG1 Isotype: IgG1 Clone: AD1.1.10 Dye: DyLight 650 Excitation max. λ: 652 nm Emission max. λ: 672 nm Uses: Flow cytometry, HTS, FRET
Catalog No. 50-295-1353