Learn More
American Research Products Inc Mouse anti double stranded DNA
Supplier:  American Research Products Inc CSBE12911M
Encompass
Mouse anti-double stranded DNA antibody (IgM) ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma Method Capture ELISA
Catalog No. 50-003-29841