Learn More
Creative Biolabs Mouse Anti-Human Complement C1q Monoclonal Antibody, TJL-03, HRP conjugated, suitable for WB, IHC, ELISA, storage: dark at 4°C, freeze-thaw: do not freeze

Supplier: Creative Biolabs CTMM0322JL15
This HRP-conjugated mouse monoclonal antibody is specifically developed for detecting human Complement C1q, known as TJL-03. It offers excellent specificity and affinity, making it ideal for various applications such as Western Blot, Immunohistochemistry, and ELISA. With a proven track record of stability and reliable results, this antibody minimizes background noise to provide accurate findings.
- Target: Human Complement C1q
- Host species: Mouse
- Clone name: TJL-03
- Conjugation: HRP
- Reactivity: Human
- Applications: WB, IHC, ELISA
- Product type: Monoclonal antibody
- Storage condition: Store in dark at 4°C
- Freeze-thaw: Do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.