Learn More
Medchemexpress LLC Mouse Anti-Human IgG Fc | 5 MG

Supplier: Medchemexpress LLC HYP837275MG
Mouse Anti-Human IgG Fc is a mouse-derived, non-conjugated IgG2a recombinant monoclonal antibody specifically targeting IgG Fc. It is expressed from CHO cells, with a heavy chain type of mIgG2a and a light chain type of mkappa. This product is intended for research use only.
- Mouse-derived, non-conjugated IgG2a recombinant monoclonal antibody
- Targets IgG Fc
- Expressed from CHO cells
- Predicted band size: 147.4 kDa
- Affinity purified
- Supplied in 0.01M phosphate buffered saline, pH 7.2, containing 1% BSA, 50% glycerol, and 0.05% sodium azide
- Stored at -20°C for 1 year, protected from light
- Avoid repeated freeze/thaw cycles
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.