Learn More
American Research Products Inc Mouse Anti IL6 Monoclonal Ab
Supplier:  American Research Products Inc IVMB03635MG
Encompass
Mouse Anti IL-6 Monoclonal Antibody In Vivo Antibody - Low Endotoxin is a in vivo antibody with mouse reactivity produced in rat Available in 1 mg size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles
Catalog No. 50-003-04900