Learn More
Santa Cruz Biotechnology MOUSE ANTI-RAB IGG HRP CONJ
Supplier:  Santa Cruz Biotechnology SC2357
Encompass
mouse anti-rabbit IgG HRP, Conventional Secondary Antibodies for Western Blotting, supplied at 200microgram in 0.5 ml volume, to be used in Western blotting at a dilution of 1:500 - 1:10000
Catalog No. NC9881816