Learn More
Sartorius Mouse Antibody Internalization
Supplier:  Sartorius 90567
Encompass
No wash protocol- fast easy antibody-labeling method optimal for screening antibody internalizationHigh throughput evaluation of antibody internalization- Assay designed to measure internalization of a large number of antibodies in 96- and 384-well plate formats using 20 L of cells and antibody.Cell viability can also be measured by multiplexing iQue Cell Membrane Integrity Dye to assess general cell health and antibody function.Cell specificity can be characterized by multiplexing Intellicyt Encoding dyes cell lines or directly conjugated fluorescent antibodies cells .The Qbeads Assay can also be multiplexed to perform more detailed antibody assessments on the sample such as cytokine release.Available in Human and Mouse formats
Catalog No. 50-289-1592