Learn More
American Research Products Inc Mouse AntiCD11c In Vivo Ab
Supplier:  American Research Products Inc IVMB028550UG
Encompass
Mouse Anti-CD11c Low Endotoxin In Vivo Antibody (Phenocycler-Fusion Validated) is a in vivo antibody with mouse reactivity produced in armenian hamster Available in 50 ug size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles
Catalog No. 50-003-06990