Learn More
American Research Products Inc Mouse antidiuretic hormone Vas
Supplier:  American Research Products Inc CSBE09272M
Encompass
Mouse antidiuretic hormone/Vasopressin/arginine Vasopressin ADH/VP/AVP ELISA Kit from CUSABIO Detection Range 1 25 pg/mL-80 pg/mL Sensitivity 0 31 pg/mL Sample serum plasma cell culture supernates Method Sandwich
Catalog No. 50-003-28679