Learn More
American Research Products Inc Mouse AntiPD1
Supplier:  American Research Products Inc IVMB0281100MG
Encompass
Mouse Anti-PD-1 (CD279) [RMP1-14] Recombinant Monoclonal Antibody is a in vivo antibody with mouse reactivity produced in mouse-rat chimeric Available in 100 mg size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles
Catalog No. 50-003-06481
Only null left
Add to Cart
Request a Quote