Learn More
Cayman Chemical Mouse Aryl Hydrocarbon Recept
SDP
Supplier:  Cayman Chemical 3690496 WELL
Encompass_Preferred
INDIGO’s Mouse AhR assay kit is an all-inclusive system. In addition to Mouse AhR Reporter Cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, the reference agonist MeBIO, Luciferase Detection Reagent, and a cell culture-ready assay plate.
Catalog No. 50-291-5188