Learn More
American Research Products Inc Mouse Aspartate Aminotransfera
Supplier:  American Research Products Inc CSBE12649M
Encompass
Mouse Aspartate Aminotransferase (AST) ELISA kit from CUSABIO Detection Range 1 56 mIU/mL-100 mIU/mL Sensitivity 0 39 mIU/mL Sample serum plasma cell culture supernates Method Sandwich
Catalog No. 50-003-29542