Learn More
American Research Products Inc Mouse B and T lymphocyte atte
Supplier:  American Research Products Inc CSBEL002868MO
Encompass
Mouse B- and T-lymphocyte attenuator (BTLA) ELISA kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27044