Learn More
Lonza Walkersville Mouse B6C3F1 Heps Cryo Suspens
Supplier:  Lonza Walkersville MXCS01
Encompass
All our suspension cryopreserved primary animal hepatocytes are specifically pre-tested for metabolic activity of general Phase I and Phase II metabolism in suspension They are not suitable for applications requiring adherent cells
Catalog No. 50-004-96250