Learn More
ACROBiosystems ACROBiosystems Mouse B7-2 / CD86 Protein, Fc Tag (MALS verified)
Supplier:  ACROBiosystems CD6M5251100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse B7-2, Fc Tag (CD6-M5251) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Glu 245 (Accession # NP_062261.3).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.9 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse B7-2, Fc Tag (Cat. No. CD6-M5251) at 5 μg/mL (100 μL/well) can bind Mouse CTLA-4, His Tag (Cat. No. CT4-M52H5) with a linear range of 0.4-13 ng/mL (QC tested).;Immobilized Biotinylated Mouse CD28, His,Avitag (Cat. No. CD8-M82E3) at 1 μg/mL (100 μL/well) on Streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Mouse B7-2, Fc Tag (Cat. No. CD6-M5251) with a linear range of 2-39 ng/mL (Routinely tested).
Catalog No. 50-210-6566