Learn More
Vector Biolabs mouse BECN1 shRNA Adenovirus (Ad-m-BECN1-shRNA)
Supplier:  Vector Biolabs 1811
Encompass
This is an pre-made gene silencing adenovirus that expresses a shRNA to knockdown mouse BECN1 gene. The shRNA expression is driven by an U6 promoter.
The knockdown of this mouse gene was validated by western blot in Ht22 cells.
Catalog No. 50-004-96054