Learn More
Santa Cruz Biotechnology MOUSE BETA ACTIN ANTIBODY
Supplier:  Santa Cruz Biotechnology SC47778
Encompass
Anti-beta-Actin (C4); Source: Mouse; Antibody Type: Monoclonal; Isotype: IgG1; Species Reactivity: Mouse, Rat, Human, Rabbit, Dictyostelium discoideum and Physarum polycephalum; 200ug/mL; Tested applications: WB, IP, IF and IHC(P); 200ug/mL
Catalog No. NC9464046