Learn More
Raybiotech Inc Mouse BNP ELISA Kit
Supplier:  Raybiotech Inc EIAMBNP1
Encompass_Preferred
RayBio Mouse/Rat Brain Natriuretic Peptide EIA Kit optimized for serum and cell culture medium. Competition-based ELISA on a 96-well strip plate.
Catalog No. NC1784165