Learn More
American Research Products Inc Mouse Brain deriVed neurotroph
Supplier:  American Research Products Inc CSBE04505M
Encompass
Mouse Brain deriVed neurotrophic factor BDNF ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26380