Learn More
American Research Products Inc Mouse Calcium dependent phosph
Supplier:  American Research Products Inc CSBEL018103MO
Encompass
Mouse Calcium-dependent phospholipase A2 (PLA2G5) ELISA kit from CUSABIO Detection Range 62 5 pg/ml-4000 pg/ml Sensitivity 15 6 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28029