Learn More
American Research Products Inc Mouse carcinoembryonic antign
Supplier:  American Research Products Inc CSBE13925M
Encompass
Mouse carcinoembryonic antign CEA ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29400