Learn More
Sino Biological Mouse CCL2 ELISA Pair Set 15PP (Sino Biological)
Supplier:  Sino Biological SEK5036815
Encompass_Preferred
Each vial contains 28 ng of recombinant Mouse MCP-1/CCL2. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 600 pg/mL is recommended.
Catalog No. 50-237-0478