Learn More
Bon Opus Biosciences Recombinant Mouse CCL22 (C-mFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP108750UG
Encompass_Preferred
Recombinant Mouse CCL22 (C-mFc) is produced by Human Cells. The target gene encoding Gly25-Ser92 is expressed with a C-mFc tag. Accession# NP_033163.1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1087
Catalog No. 50-217-2685