Learn More
Zyagen Labs Mouse CD1 Spleen Total Protein, 1mg
SDP
Supplier:  Zyagen Labs MT701
Encompass_Preferred
Mouse Spleen total protein (protein lysate) is isolated from freshly harvested tissues of single healthy normal donor. Tissues are homogenized in protein lysis buffer supplemented with a cocktail of 6 mammalian protease inhibitors to minimize proteolysis, precisely quantified, and stored at -80oC.
Catalog No. NC3585205