Learn More
Sino Biological Anti-CD200 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50074RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD200 (rM CD200; Catalog#50074-M08H; O54901.1; Met 1-Gly 232). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3480