Learn More
ACROBiosystems Mouse CD206 25ug
Supplier:  ACROBiosystems CD6M82E825UG
Encompass_Preferred
Biotinylated Mouse CD206 Protein HisAvitag (CD6-M82E8) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Ala 1388 (Accession # Q61830).
Catalog No. 50-253-5354