Learn More
ACROBiosystems ACROBiosystems Mouse CD27 Ligand / CD70 Protein, His Tag (active trimer) (MALS verified)
Supplier:  ACROBiosystems CDLM5245100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse CD27 Ligand, His Tag (active trimer) (MALS verified) (CDL-M5245) is expressed from human 293 cells (HEK293). It contains AA Gln 47 - Pro 195 (Accession # O55237-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 18.6 kDa. The protein migrates as 27-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse CD27, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD7-M5252) at 2 μg/mL (100 μL/well) can bind Mouse CD27 Ligand, His Tag (active trimer) (MALS verified) (Cat. No. CDL-M5245) with a linear range of 0.1-4 ng/mL (QC tested).
Catalog No. 50-210-6589