Learn More
American Research Products Inc Mouse choline acetyltransteras
Supplier:  American Research Products Inc CSBE17524M
Encompass
Mouse choline acetyltransterase (ChAT) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates cell culture supernates Method Sandwich
Catalog No. 50-003-28461