Learn More
Sino Biological Recombinant Mouse Claudin18 Protein (CLDN18.2, Full-Length, His & Strep Tag), VLP
Supplier:  Sino Biological 59437M64H100
Encompass_Preferred
A DNA sequence encoding the Mouse CLDN18 (Full Length) (P56857-3) (Ser2-Val264) was expressed, with a polyhistidine tag at the N-terminus and a Strep tag at the C-terminus.
Catalog No. 50-299-3162