Learn More
Sino Biological Recombinant Mouse Claudin-6 Protein (VLP, Full-length, His & FLAG Tag)
Supplier:  Sino Biological 59110M61H100
Encompass_Preferred
A DNA sequence encoding the Mouse CLDN6 (Full Length) (Q9Z262) (Met1-Val219) was expressed, with a polyhistidine tag followed by a Flag tag at the C-terminus.
Catalog No. 50-299-2733