Learn More
BIOHIPPO Mouse CNP Microsample
SDP
Supplier:  BIOHIPPO BHE15210744
Encompass
Mouse CNP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CNP in serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 0 95 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CNP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CNP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CNP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14601