Learn More
American Research Products Inc Mouse coagulation factor III
Supplier:  American Research Products Inc CSBE17813M
Encompass
Mouse coagulation factor III FIII ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27746
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.